1-Iodoperfluorodecane ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: Henicosafluoro-10-iododecane, Perfluorodecyl iodide, Perfluoro-1-iododecane

003131-5G 003131-25G
101785-292EA 60 USD
101785-292 101785-294
1-Iodoperfluorodecane ≥97%
1-Iodoperfluorodecane

Hazard code(s): Irritant

Formula: C₁₀F₂₁I
MW: 645.98 g/mol
Boiling Pt: 195…200 °C
Melting Pt: 65…67 °C
MDL Number: MFCD00001065
CAS Number: 423-62-1
EINECS: 207-030-5

Order Now

Specification Test Results

Boiling point 195-200°C
Melting point 65-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR