Methyl-2-cyanobenzoate ≥99%

Supplier: Matrix Scientific

003120-1G 003120-5G
101785-274EA 56.92 USD
101785-274 101785-276
Methyl-2-cyanobenzoate ≥99%
Methyl-2-cyanobenzoate

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₉H₇NO₂
Boiling Pt: 108 °C (1mm)
Melting Pt: 49…51 °C
MDL Number: MFCD00016376
CAS Number: 6587-24-2

Order Now

Specification Test Results

Boiling point 108°C/1mm
Melting point 49-51°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR