2-(2,4-Dichlorophenoxy)ethanol
Supplier: Matrix Scientific
003099-5G
003099-25G
101785-238EA
100
USD
101785-238
101785-240
2-(2,4-Dichlorophenoxy)ethanol
2-(2,4-Dichlorophenoxy)ethanol
Hazard code(s): Irritant
Formula:
C₈H₈Cl₂O₂ Melting Pt: 54 °C |
MDL Number:
MFCD00016564 CAS Number: 120-67-2 EINECS: 204-416-5 |
Specification Test Results
Melting point | 54°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...