3-(3,4-Dichlorophenyl)propionic acid

Supplier: Matrix Scientific

003098-1G 003098-5G
101785-234EA 26.15 USD
101785-234 101785-236
3-(3,4-Dichlorophenyl)propionic acid
3-(3,4-Dichlorophenyl)propionic acid

Hazard code(s): Irritant

Formula: C₉H₈Cl₂O₂
Melting Pt: 97…99 °C
MDL Number: MFCD00016551
CAS Number: 25173-68-6

Order Now

Specification Test Results

Melting point 97-99°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR