3-(2,4-Dichlorophenyl)propionic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
3-(2,4-Dichlorophenyl)propanoic acid
003096-5G
003096-25G
101785-226EA
56.92
USD
101785-226
101785-228
3-(2,4-Dichlorophenyl)propionic acid ≥97%
3-(2,4-Dichlorophenyl)propionic acid
Hazard code(s): Irritant
Formula:
C₉H₈Cl₂O₂ MW: 219.07 g/mol Melting Pt: 89…93 °C |
MDL Number:
MFCD00275605 CAS Number: 55144-92-8 |
Specification Test Results
Melting point | 90-90°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...