3,4,5-Trichloro-α,α,α-trifluorotoluene ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 3,4,5-Trichlorobenzotrifluoride, 1,2,3-Trichloro-5-(trifluoromethyl)benzene

003093-25G 003093-100G
101785-222EA 35.38 USD
101785-222 101785-224
3,4,5-Trichloro-α,α,α-trifluorotoluene ≥97% for synthesis
3,4,5-Trichloro-α,α,α-trifluorotoluene

Hazard code(s): Irritant

Formula: C₇H₂Cl₃F₃
Boiling Pt: 113 °C (50 -rr)
Melting Pt: -1 °C
Density: 1.6003 g/cm³ (20 °C)
MDL Number: MFCD00035985
CAS Number: 50594-82-6
EINECS: 256-636-6
UN: 2234
ADR: 3,III

Order Now

Specification Test Results

Boiling point 200-206°C
Density 1.6
ND20 1.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR