2,4,6-Trichlorobenzonitrile ≥98%

Supplier: Matrix Scientific

003092-5G 003092-25G
101785-218EA 87.69 USD
101785-218 101785-220
2,4,6-Trichlorobenzonitrile ≥98%
2,4,6-Trichlorobenzonitrile

Hazard code(s): Toxic

Formula: C₇H₂Cl₃N
Melting Pt: 80…82 °C
MDL Number: MFCD00052711
CAS Number: 6575-05-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 74-76°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR