Methyl-2-chloro-4-nitrobenzoate ≥99%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-4-nitrobenzoic acid methyl ester, Methyl 2-chloro-4-nitrobenzoate
003089-5G
003089-25G
101785-204EA
44.62
USD
101785-204
101785-206
Methyl-2-chloro-4-nitrobenzoate ≥99%
Methyl-2-chloro-4-nitrobenzoate
Hazard code(s): Irritant
Formula:
C₈H₆ClNO₄ MW: 215.59 g/mol Melting Pt: 72…75 °C |
MDL Number:
MFCD00017013 CAS Number: 13324-11-3 EINECS: 236-362-3 |
Specification Test Results
Melting point | 72-75°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...