Methyl-2-chloro-4-fluorobenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloro-4-fluorobenzenecarboxylate, Methyl 2-chloro-4-fluorobenzoate, 2-Chloro-4-fluorobenzoic acid methyl ester

003088-5G 003088-25G 003088-100G
101785-198EA 16.92 USD
101785-198 101785-200 101785-202
Methyl-2-chloro-4-fluorobenzoate ≥97%
Methyl-2-chloro-4-fluorobenzoate

Hazard code(s): Irritant

Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
Boiling Pt: 103…105 °C (15mm)
MDL Number: MFCD00173937
CAS Number: 85953-29-3

Order Now

Specification Test Results

Boiling point 103-105°C/1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR