1,1,2-Trichloro-2,3,3-trifluorocyclobutane ≥97%
Supplier: Matrix Scientific
003082-5G
003082-25G
101785-178EA
76.92
USD
101785-178
101785-180
1,1,2-Trichloro-2,3,3-trifluorocyclobutane ≥97%
1,1,2-Trichloro-2,3,3-trifluorocyclobutane
Hazard code(s): Irritant
Formula:
C₄H₂Cl₃F₃ MW: 213.41 g/mol Boiling Pt: 120 °C Melting Pt: –32 °C Density: 1.59 g/cm³ (20 °C) |
MDL Number:
MFCD00039439 CAS Number: 697-17-6 EINECS: 211-804-8 |
Specification Test Results
Boiling point | 120°C |
Density | 1.594 |
ND20 | 1.4163 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...