3-Acetylbenzonitrile ≥98%

Supplier: Matrix Scientific

003069-5G 003069-25G
101785-154EA 21.54 USD
101785-154 101785-156
3-Acetylbenzonitrile ≥98%
3-Acetylbenzonitrile

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₉H₇NO
Boiling Pt: 120 °C (5 -rr)
Melting Pt: 98 - 100 °C
MDL Number: MFCD00001806
CAS Number: 6136-68-1
EINECS: 228-110-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 119°C/1mm
Melting point 99-100°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR