2,3,4-Tribromothiophene ≥97%

Supplier: Matrix Scientific

003046-1G 003046-5G
101785-132EA 89.23 USD
101785-132 101785-134
2,3,4-Tribromothiophene ≥97%
2,3,4-Tribromothiophene

Hazard code(s): Irritant

Formula: C₄HBr₃S
MW: 320.83 g/mol
Boiling Pt: 132 - 134 °C (11 -rr)
Melting Pt: 43 - 46 °C
MDL Number: MFCD00041210
CAS Number: 3141-25-1
EINECS: 221-545-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 43-45°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR