α,α,α-Trifluoro-o-cresol ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxybenzotrifluoride, 2-(Trifluoromethyl)phenol

003024-25G 003024-100G
101785-102EA 61.54 USD
101785-102 101785-104
α,α,α-Trifluoro-o-cresol ≥99%
α,α,α-Trifluoro-o-cresol

Hazard code(s): Corrosive

Formula: C₇H₅F₃O
MW: 162.11 g/mol
Boiling Pt: 147…148 °C
Melting Pt: 45…46 °C
Density: Min. 1 g/cm³ (20 °C)
MDL Number: MFCD00002222
CAS Number: 444-30-4
EINECS: 207-148-7
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Boiling point 147-148°C
Melting point 45-46°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR