7-Methoxybenzofuran ≥90%
Supplier: Matrix Scientific
003012-1G
003012-5G
101785-066EA
110.77
USD
101785-066
101785-068
7-Methoxybenzofuran ≥90%
7-Methoxybenzofuran
Hazard code(s): Irritant
Formula:
C₉H₈O₂ MW: 148.16 g/mol |
MDL Number:
MFCD01310825 CAS Number: 7168-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...