5-Bromoisatoic anhydride ≥90% for synthesis

Supplier: Matrix Scientific

002992-1G 002992-5G
101785-020EA 20 USD
101785-020 101785-022
5-Bromoisatoic anhydride ≥90% for synthesis
5-Bromoisatoic anhydride

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₈H₄BrNO₃
Melting Pt: 285…290 °C (dec.)
MDL Number: MFCD00016921
CAS Number: 4692-98-2

Order Now

Specification Test Results

Melting point 285-290°C(d


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR