3-Fluoropyrocatechol ≥99%

Supplier: Matrix Scientific

Synonyms: 3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene

002989-1G 002989-5G
101785-012EA 33.85 USD
101785-012 101785-014
3-Fluoropyrocatechol ≥99%
3-Fluoropyrocatechol

Hazard code(s): Irritant

Formula: C₆H₅FO₂
MW: 128.1 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 71…73 °C
Density: 1.514 g/cm³ (20 °C)
MDL Number: MFCD00042582
CAS Number: 363-52-0

Order Now

Specification Test Results

Melting point 71-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR