(±)-4-Fluoromandelic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-Fluoromandelic Acid, (±)-4-Fluorophenylglycolic acid

002973-5G 002973-25G 002973-100G
101784-988EA 21.54 USD
101784-988 101784-990 101784-992
(±)-4-Fluoromandelic acid ≥98% for synthesis
(±)-4-Fluoromandelic acid

Hazard code(s): Irritant

Formula: C₈H₇FO₃
Melting Pt: 136…137 °C
MDL Number: MFCD00044402
CAS Number: 395-33-5
EINECS: 206-899-8

Order Now

Specification Test Results

Melting point 136-137°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR