2-Chloro-4'-fluoroacetanilide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-N-(4-fluorophenyl)acetamide, N-(Chloroacetyl)-4-fluoroaniline

002928-5G 002928-25G 002928-100G
101784-936EA 21.54 USD
101784-936 101784-938 101784-940
2-Chloro-4'-fluoroacetanilide ≥97%
2-Chloro-4'-fluoroacetanilide

Hazard code(s): Irritant

Formula: C₈H₇ClFNO
MW: 187.6 g/mol
MDL Number: MFCD00015730
CAS Number: 351-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR