2'-Ethoxyacetophenone ≥99%

Supplier: Matrix Scientific

Synonyms: 1-(2-Ethoxyphenyl)ethanone, 1-(2-Ethoxyphenyl)ethan-1-one

002920-5G 002920-25G
101784-910EA 27.69 USD
101784-910 101784-912
2'-Ethoxyacetophenone ≥99%
2'-Ethoxyacetophenone

Hazard code(s): Irritant

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 243.5 °C (1013 hPa)
Melting Pt: 36 - 40 °C
Density: 1.0036 g/cm³ (78 °C)
MDL Number: MFCD00017262
CAS Number: 2142-67-8

Order Now

Specification Test Results

Boiling point 243-244°C
Melting point 43°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR