4'-Phenoxyacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: p-Phenoxyacetophenone

002918-5G 002918-25G
101784-900EA 41.54 USD
101784-900 101784-902
4'-Phenoxyacetophenone ≥98%
4'-Phenoxyacetophenone

Hazard code(s): Irritant

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 119…120 °C (0.03 mmHg)
Melting Pt: 50…52 °C
MDL Number: MFCD00008744
CAS Number: 5031-78-7

Order Now

Specification Test Results

Melting point 50-52°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR