2'-Fluoroacetanilide ≥98%
Supplier: Matrix Scientific
002906-25G
002906-100G
101784-888EA
35.38
USD
101784-888
101784-890
2'-Fluoroacetanilide ≥98%
2'-Fluoroacetanilide
MDL: MFCD00000315
Formula:
C₈H₈FNO MW: 153.16 g/mol Boiling Pt: 140 - 142 °C (14 -rr) Melting Pt: 81 °C |
MDL Number:
MFCD00000315 CAS Number: 399-31-5 EINECS: 206-916-9 |
Specification Test Results
Boiling point | 140-142°C/1 |
Melting point | 76-78°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...