2',4',6'-Trifluoropropiophenone ≥97%

Supplier: Matrix Scientific

002889-1G 002889-5G
101784-864EA 130.77 USD
101784-864 101784-866
2',4',6'-Trifluoropropiophenone ≥97%
2',4',6'-Trifluoropropiophenone

Hazard code(s): Irritant

Formula: C₉H₇F₃O
MW: 188.15
Boiling Pt: 67…69 °C (3mm)
MDL Number: MFCD00061224
CAS Number: 220141-69-5

Order Now

Specification Test Results

Boiling point 67-69°C/3mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR