3',4',5'-Trifluoroacetophenone

Supplier: Matrix Scientific

Synonyms: 1-(3,4,5-Trifluorophenyl)ethan-1-one

002876-1G 002876-5G
101784-820EA 30.77 USD
101784-820 101784-822
3',4',5'-Trifluoroacetophenone
3',4',5'-Trifluoroacetophenone
Formula: C₈H₅F₃O
MW: 174.12 g/mol
MDL Number: MFCD00083533
CAS Number: 220141-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR