2',3',6'-Trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

002874-1G 002874-5G
101784-810EA 24.62 USD
101784-810 101784-812
2',3',6'-Trifluoroacetophenone ≥97%
2',3',6'-Trifluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₅F₃O
MDL Number: MFCD00070809
CAS Number: 208173-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR