1H,1H,2H-Perfluoro-1-dodecene ≥96%

Supplier: Matrix Scientific

002864-5G 002864-25G 002864-100G
101784-796EA 46.15 USD
101784-796 101784-798 102550-824
1H,1H,2H-Perfluoro-1-dodecene ≥96%
1H,1H,2H-Perfluoro-1-dodecene

Hazard code(s): Irritant, Refrigerate

Formula: C₁₂H₃F₂₁
Boiling Pt: 71…72 °C (14mm)
Density: 1.711 g/cm³
MDL Number: MFCD00042346
CAS Number: 30389-25-4
EINECS: 250-173-3

Order Now

Specification Test Results

Boiling point 71-72°C/14m
Density 1.711


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR