1H,1H,9H-Hexadecafluoro-1-nonanol ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1-nonanol, 1H,1H,9H-Perfluoro-1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluor-1-nonanol, 1H,1h,9h-hexadecafluorononanol

002834-25G 002834-100G
101784-750EA 24.62 USD
101784-750 101784-752
1H,1H,9H-Hexadecafluoro-1-nonanol ≥97% for synthesis
1H,1H,9H-Hexadecafluoro-1-nonanol

Hazard code(s): Irritant

Formula: C₉H₄F₁₆O
Boiling Pt: 157…158 °C (200mm)
Melting Pt: 60…65 °C
MDL Number: MFCD00039629
CAS Number: 376-18-1
EINECS: 206-806-0

Order Now

Specification Test Results

Boiling point 157-158°C/2
Melting point 60-65°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR