2-Acetyl-7-methoxybenzofuran ≥97%

Supplier: Matrix Scientific

002805-1G 002805-5G
101784-698EA 26.15 USD
101784-698 101784-700
2-Acetyl-7-methoxybenzofuran ≥97%
2-Acetyl-7-methoxybenzofuran

Hazard code(s): Irritant

Formula: C₁₁H₁₀O₃
Melting Pt: 94…95 °C
MDL Number: MFCD00016911
CAS Number: 43071-52-9

Order Now

Specification Test Results

Melting point 94-95°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR