2-Fluoro-6-phenoxybenzonitrile ≥95%
Supplier: Matrix Scientific
002798-5G
002798-25G
101784-688EA
36.92
USD
101784-688
101784-690
2-Fluoro-6-phenoxybenzonitrile ≥95%
2-Fluoro-6-phenoxybenzonitrile
Hazard code(s): Irritant
Formula:
C₁₃H₈FNO MW: 213.21 g/mol Boiling Pt: 338 °C (1013 hPa) Melting Pt: 90…93 °C |
MDL Number:
MFCD00068201 CAS Number: 175204-06-5 |
Specification Test Results
Melting point | 90-93°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...