2-Chloro-6-iodotoluene ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Chloro-3-iodo-2-methylbenzene

002788-25G 002788-100G
101784-664EA 47.69 USD
101784-664 101784-666
2-Chloro-6-iodotoluene ≥99%
2-Chloro-6-iodotoluene

Hazard code(s): Irritant

Formula: C₇H₆ClI
MW: 252.48 g/mol
Boiling Pt: 117 °C (20mm)
Density: 1.82 g/cm³
MDL Number: MFCD00019013
CAS Number: 42048-11-3

Order Now

Specification Test Results

Boiling point 117°C/20mm
Density 1.82

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR