2-Fluoro-6-iodobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

002785-5G 002785-25G
101784-656EA 44.62 USD
101784-656 101784-658
2-Fluoro-6-iodobenzoic acid ≥98%
2-Fluoro-6-iodobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄FIO₂
MW: 266.01 g/mol
Boiling Pt: 312 °C (1013 hPa)
Melting Pt: 118…120 °C
MDL Number: MFCD00042289
CAS Number: 111771-08-5

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR