6-Fluorosalicylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Fluoro-2-hydroxybenzoic acid, 2-Fluoro-6-hydroxybenzoic acid
002783-1G
002783-5G
002783-25G
101784-646EA
26.15
USD
101784-646
101784-648
101784-650
6-Fluorosalicylic acid ≥97%
6-Fluorosalicylic acid
Hazard code(s): Irritant
Formula:
C₇H₅FO₃ MW: 156.11 g/mol Melting Pt: 159…161 °C |
MDL Number:
MFCD00052179 CAS Number: 67531-86-6 |
Specification Test Results
Melting point | 159-161°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...