(2-Chloro-6-fluorophenyl)acetic acid ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 2-Chloro-6-fluorophenylacetic acid

002777-5G 002777-25G
101784-630EA 29.23 USD
101784-630 101784-632
(2-Chloro-6-fluorophenyl)acetic acid ≥98% for synthesis
(2-Chloro-6-fluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃ClF)CH₂COOH
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 120 - 123 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004319
CAS Number: 37777-76-7
EINECS: 253-661-4

Order Now

Specification Test Results

Melting point 120-123°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR