2-Chloro-6-fluorobenzaldehyde ≥97%

Supplier: Matrix Scientific
Warning

002774-100G 002774-500G
101784-626EA 38.46 USD
101784-626 101784-628
2-Chloro-6-fluorobenzaldehyde ≥97%
2-Chloro-6-fluorobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₄ClFO
Boiling Pt: 91…93 °C (10mm)
Melting Pt: 35…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00003306
CAS Number: 387-45-1
EINECS: 206-860-5

Order Now

Specification Test Results

Boiling point 91-93°C/10m
Melting point 35-38°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR