2-Chloro-6-fluorobenzoic acid ≥97%

Supplier: Matrix Scientific
Warning

002766-25G 002766-100G
101784-614EA 21.54 USD
101784-614 101784-616
2-Chloro-6-fluorobenzoic acid ≥97%
2-Chloro-6-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄ClFO₂
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00002417
CAS Number: 434-75-3
EINECS: 207-105-2

Order Now

Specification Test Results

Melting point 159-161°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR