Methyl-2,6-difluorobenzoate ≥98%
Supplier: Matrix Scientific
002748-25G
002748-100G
101784-572EA
33.85
USD
101784-572
101784-574
Methyl-2,6-difluorobenzoate ≥98%
Methyl-2,6-difluorobenzoate
Hazard code(s): Irritant
Formula:
C₈H₆F₂O₂ Boiling Pt: 80…82 °C (15mm) |
MDL Number:
MFCD00051778 CAS Number: 13671-00-6 |
Specification Test Results
Boiling point | 80-82°C/15m |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...