2-Fluoro-6-(trifluoromethyl)benzyl alcohol ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: (2-Fluoro-6-(trifluoromethyl)phenyl)methanol

002742-1G 002742-5G
101784-560EA 30.77 USD
101784-560 101784-562
2-Fluoro-6-(trifluoromethyl)benzyl alcohol ≥97% for synthesis
2-Fluoro-6-(trifluoromethyl)benzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₆F₄O
MDL Number: MFCD00061168
CAS Number: 152211-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR