2-Fluoro-6-(trifluoromethyl)benzaldehyde ≥97%

Supplier: Matrix Scientific

002739-1G 002739-5G
101784-556EA 33.85 USD
101784-556 101784-558
2-Fluoro-6-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-6-(trifluoromethyl)benzaldehyde

Hazard code(s): Irritant

Formula: C₈H₄F₄O
MW: 192.11 g/mol
MDL Number: MFCD00061273
CAS Number: 60611-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR