2-Chloro-5-nitrobenzotrifluoride ≥97%

Supplier: Matrix Scientific

002720-25G 002720-100G
101784-526EA 24.62 USD
101784-526 101784-528
2-Chloro-5-nitrobenzotrifluoride ≥97%
2-Chloro-5-nitrobenzotrifluoride

Hazard code(s): Irritant

Formula: C₇H₃ClF₃NO₂
Boiling Pt: 102…103 °C (5mm)
Melting Pt: 25…27 °C
Density: 1.527 g/cm³
MDL Number: MFCD00007296
CAS Number: 777-37-7
EINECS: 212-287-1

Order Now

Specification Test Results

Boiling point 102-103°C/5
Density 1.527
ND20 1.508


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR