2-Fluoro-5-nitrobenzaldehyde ≥97%

Supplier: Matrix Scientific

002718-5G 002718-25G 002718-100G
101784-514EA 18.46 USD
101784-514 101784-516 101784-518
2-Fluoro-5-nitrobenzaldehyde ≥97%
2-Fluoro-5-nitrobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₄FNO₃
MW: 169.11 g/mol
Melting Pt: 57…60 °C
MDL Number: MFCD00042298
CAS Number: 27996-87-8

Order Now

Specification Test Results

Melting point 57-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR