2-Fluoro-5-nitrobenzoic acid ≥97%

Supplier: Matrix Scientific
Warning

002716-5G 002716-25G 002716-100G
101784-508EA 15.38 USD
101784-508 101784-510 101784-512
2-Fluoro-5-nitrobenzoic acid ≥97%
2-Fluoro-5-nitrobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄FNO₄
Melting Pt: 142…144 °C
MDL Number: MFCD00134238
CAS Number: 7304-32-7

Order Now

Specification Test Results

Melting point 142-144°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR