2-Fluoro-5-nitroaniline ≥98%

Supplier: Matrix Scientific

002713-25G 002713-100G
101784-504EA 21.54 USD
101784-504 101784-506
2-Fluoro-5-nitroaniline ≥98%
2-Fluoro-5-nitroaniline

Hazard code(s): Toxic

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00007652
CAS Number: 369-36-8
EINECS: 206-720-3
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR