6-Fluoro-m-cresol ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 2-Fluoro-5-methylphenol, 1-Fluoro-2-hydroxy-4-methylbenzene

002710-5G 002710-25G 002710-100G
101784-498EA 23.08 USD
101784-498 101784-500 101784-502
6-Fluoro-m-cresol ≥97%
6-Fluoro-m-cresol

Hazard code(s): Irritant

Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 173 °C
Density: 1.155 g/cm³
MDL Number: MFCD00190101
CAS Number: 63762-79-8
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Boiling point 173°C
Density 1.155
ND20 1.511

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR