6-Chloro-m-toluic acid ≥98%

Supplier: Matrix Scientific
Warning

002709-1G 002709-5G 002709-25G
101784-492EA 30.77 USD
101784-492 101784-494 101784-496
6-Chloro-m-toluic acid ≥98%
6-Chloro-m-toluic acid

Hazard code(s): Irritant

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Melting Pt: 149…151 °C
MDL Number: MFCD00045798
CAS Number: 6342-60-5

Order Now

Specification Test Results

Melting point 149-151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR