2-Chloro-5-iodobenzoic acid ≥98%

Supplier: Matrix Scientific

002704-25G 002704-100G
101784-476EA 52.31 USD
101784-476 101784-478
2-Chloro-5-iodobenzoic acid ≥98%
2-Chloro-5-iodobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄ClIO₂
Melting Pt: 161…163 °C
MDL Number: MFCD00079731
CAS Number: 19094-56-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 161-163°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR