4-Chloro-3-(trifluoromethyl)phenol ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-hydroxybenzotrifluoride

002702-5G 002702-25G
101784-466EA 41.54 USD
101784-466 101784-468
4-Chloro-3-(trifluoromethyl)phenol ≥98%
4-Chloro-3-(trifluoromethyl)phenol

Hazard code(s): Irritant

Formula: C₇H₄ClF₃O
MW: 196.56 g/mol
MDL Number: MFCD00042525
CAS Number: 6294-93-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR