2-Chloro-5-fluorobenzyl alcohol ≥97%

Supplier: Matrix Scientific

002698-5G 002698-25G
101784-462EA 52.31 USD
101784-462 101784-464
2-Chloro-5-fluorobenzyl alcohol ≥97%
2-Chloro-5-fluorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₆ClFO
MW: 160.58 g/mol
Melting Pt: 68…72 °C
MDL Number: MFCD01631418
CAS Number: 261762-59-8

Order Now

Specification Test Results

Melting point 68-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR