3-Chloro-5-fluorobenzoic acid ≥97%

Supplier: Matrix Scientific
Warning

002695-5G 002695-25G
101784-454EA 26.15 USD
101784-454 101784-456
3-Chloro-5-fluorobenzoic acid ≥97%
3-Chloro-5-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄ClFO₂
Boiling Pt: 274.9 °C
Density: 1.477 g/cm³
MDL Number: MFCD00973904
CAS Number: 25026-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR