Methyl-2,5-dimethoxybenzoate ≥98%
Supplier: Matrix Scientific
002687-1G
002687-5G
101784-444EA
160
USD
101784-444
101784-446
Methyl-2,5-dimethoxybenzoate ≥98%
Methyl-2,5-dimethoxybenzoate
Hazard code(s): Irritant
Formula:
C₁₀H₁₂O₄ Boiling Pt: 100 °C (1mm) Density: 1.19 g/cm³ |
MDL Number:
MFCD00060624 CAS Number: 2150-40-5 |
Specification Test Results
Boiling point | 100°C/1mm |
Density | 1.19 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...