2-Acetyl-5-chlorothiophene ≥99%

Supplier: Matrix Scientific

Synonyms: 1-(5-Chloro-2-thienyl)-1-ethanone, Rivaroxaban Impurity 20

002685-25G 002685-100G
101784-440EA 123.08 USD
101784-440 101784-442
2-Acetyl-5-chlorothiophene ≥99%
2-Acetyl-5-chlorothiophene

Hazard code(s): Irritant

Formula: C₆H₅ClOS
MW: 160.62 g/mol
Boiling Pt: 117 °C (17 -rr)
Melting Pt: 52 °C
MDL Number: MFCD00005444
CAS Number: 6310-09-4
EINECS: 228-630-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 117-118°C/1
Melting point 46-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR