Dimethyl-5-bromoisophthalate ≥98%

Supplier: Matrix Scientific

Synonyms: Dimethyl 5-bromoisophthalate, 5-Bromoisophthalic acid dimethyl ester

002682-5G 002682-25G
101784-428EA 32.31 USD
101784-428 101784-430
Dimethyl-5-bromoisophthalate ≥98%
Dimethyl-5-bromoisophthalate

Hazard code(s): Irritant

Formula: C₁₀H₉BrO₄
MW: 273.08 g/mol
Melting Pt: 89…91 °C
MDL Number: MFCD00078709
CAS Number: 51760-21-5
EINECS: 257-386-0

Order Now

Specification Test Results

Melting point 89-91°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR